Learn More
CD195 Mouse anti-Human, APC-Cy7, Clone: 2D7/CCR5, BD
Mouse Monoclonal Antibody
193.67€ - 1002.59€
Spécification
| Antigene | CD195 |
|---|---|
| Clone | 2D7/CCR5 |
| Applicazioni | Flow Cytometry |
| Classificazione | Monoclonal |
| Coniugato | APC-Cyanine7 |
| Code produit | Marque | Quantità | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantità | Prix | Quantité et disponibilité | |||||
|
15870589
|
BD Biosciences
560913 |
25 Tests |
194.00€ 193.67€ / Pezzo Risparmia 0.33€ 0.17% Sconto |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
15849038
|
BD Biosciences
557755 |
100 Tests |
1005.00€ 1002.59€ / Pezzo Risparmia 2.41€ 0.24% Sconto |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Description
Reacts with the chemokine receptor, CCR5, a seven transmembrane-spanning G protein-associated molecule. CCR5 belongs to the β chemokine receptor family. It is expressed on a subset of T lymphocytes (CD3+, CD45RO+, CD95+). CCR5 regulates lymphocyte chemotaxis activation and transendothelial migration during inflammation. It signals a response to at least three chemokines: RANTES and macrophage inflammatory protein-1 (MIP-1) α and β. Additionally, CCR5 has been found to be a co-receptor for macrophage-tropic HIV-1 on CD4+ cells, a characteristic that is important in viral transmission. Reports indicate that individuals who have partial (heterozygous) or complete (homozygous) deletion of the CCR5 allele, demonstrate resistance to HIV infection. This antibody has been shown to block ligand and gp120 binding. It is also able to neutralize HIV infection. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: 2D7/CCR5
Isotype: IgG2a, κ
Species Reactivity: Human
Flow Cytometry, Functional Assay, Immunofluorescence
Spécification
| CD195 | |
| Flow Cytometry | |
| APC-Cyanine7 | |
| RUO | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| IgG2a κ | |
| Affinity Purified | |
| CCR5; C-C chemokine receptor type 5; CC-CKR-5; CKR5; CHEMR13 |
| 2D7/CCR5 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human CCR5 Transfected Cell Line | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.