missing translation for 'onlineSavingsMsg'
Learn More

CD247 with control Mouse anti-Human, Unlabeled, Clone: 8D3, BD

Mouse Monoclonal Antibody

216.14€ - 387.34€

Specifica

Antigene CD247 with control
Clone 8D3
Applicazioni Western Blot
Classificazione Monoclonal
Coniugato Unconjugated
Visualizza altre specifiche

Prodotti 2
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
15806488
Vedere i documenti
BD Biosciences
551033
50 μg
ONLINE EXCLUSIVE PRICE
217.00€
216.14€ / Pezzo
Risparmia 0.86€
0.40% Sconto
Visualizzare prodotti alternativi
Risparmia fino a 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
15816488
missing translation for 'viewDocuments'
BD Biosciences
551034
150 μg
ONLINE EXCLUSIVE PRICE
388.00€
387.34€ / Pezzo
Risparmia 0.66€
0.17% Sconto
View alternative products
missing translation for 'toSaveUpto' 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Description

Description

The T-cell receptor (TCR), expressed by thymus-derived lymphocytes, is a multi-component complex responsible for recognizing antigen in the context of MHC molecules. Ti is the antigen-specific binding component of the TCR. On the majority of peripheral T lymphocytes Ti is present as a disulfide-linked α - β heterodimer. A minor fraction express Ti as a γ- δ heterodimer. The Ti subunits are Ig-like, each containing V and C regions. Ti is noncovalently associated with an invariant set of molecules referred to as the CD3 subunits. CD3 appears early in thymocyte differentiation and remains expressed on all mature T lymphocytes. In the mouse, it consists of five chains, γ(gamma), δ (delta), ε (epsilon), ζ (zeta), and p21. The relative masses of the mouse CD3 chains are 21, 28, 26, 32 (dimer), and 21 kDa respectively.

The 8D3 antibody recognizes the ζ chain of the human and mouse T cell antigen receptor-associated CD3 complex. It reacts with 16 kDa unphosphorylated monomers and 32 kDa unphosphorylated dimers. It does not appear to recognize 21 kDa phosphorylated monomers or 42 kDa phosphorylated dimers. A synthetic peptide corresponding to a region of human CD3 ζ was used as immunogen.

Specifications

Specifications

CD247 with control
Western Blot
Unconjugated
RUO
IgG1
Affinity Purified
CD3ζ chain
8D3
Monoclonal
Mouse
Human, Mouse
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Certificates

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.