missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Human, Unlabeled, Clone: UCHT1, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 555330
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Studies from the HLDA Workshop show this antibody to be mitogenic when used in conjunction with pokeweed mitogen. CD3 plays a role in signal transduction during antigen recognition. UCHT1 antibody stains intracellular CD3 unlike the other CD3 clone, HIT3a, which stains only the extracellular CD3. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.Specifica
CD3 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human infant thymocytes and peripheral blood lymphocytes from a Sézary Syndrome donor | |
0.1 mg | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
UCHT1 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
For Research Use Only.