Learn More
anti-EGFR, CTNNA1 Protein Protein Interaction Antibody Pair, Abnova™
Marca: Abnova DI0142.1Set
Dettagli aggiuntivi : Peso : 0.00010kg
Includes: EGFR rabbit purified polyclonal antibody (20μg); CTNNA1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Descrizione
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the EGFR protein, and the other against the CTNNA1 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifica
EGFR | |
Monoclonal/Polyclonal | |
EGFR | |
EGFR | |
1 Set | |
Primary | |
Antibody pair set content: 1. EGFR rabbit purified polyclonal antibody (20 ug) 2. CTNNA1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
ERBB/ERBB1/HER1/PIG61/mENA | |
Mouse, Rabbit | |
RUO | |
1956/1495 | |
Human | |
EGFR & CTNNA1 Protein Protein Interaction Antibody Pair |