Learn More
gd T Lymphocytes Rat anti-Pig, APC, Clone: MAC320, BD
Rat Monoclonal Antibody
Marca: BD Biosciences 561482
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
The MAC320 antibody monoclonal antibody specifically binds to the 270- and 280-kDa isoforms of the SWC6 heterodimer expressed on Null T cells. These cells form a unique population of CD2-, surface Ig-, and thymus-dependent nylon wool-nonadherent peripheral blood lymphocytes found at high frequencies in young pigs. Null T cells are also CD4- and CD8-, and the majority of them express γδ T-cell receptors (TCR). In addition to peripheral blood, MAC320+ leukocytes (CD2- γδ T cells) are found in the thymus, especially in the medulla and in the marginal zone and red pulp of the spleen. They are rare in normal lymph nodes, tonsils, Peyer's patches, bone marrow, and skin. An additional population of MAC320- CD2+ γδ T cells is found in the blood and spleen. The MAC320 antibody does not react with pig erythrocytes, monocytes, or granulocytes, nor with cow, sheep, or human lymphocytes.
Specifica
gd T Lymphocytes | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Null T cells purified from the blood of young pigs | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
MAC320 | |
APC | |
Rat | |
Affinity Purified | |
RUO | |
Porcine | |
IgG2a κ |