missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gephyrin Mouse, Unlabeled, Clone: 45, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 610585
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The sub-membraneous region of the postsynaptic neuron is an intricate network of cytoskeletal elements generally known as the postsynaptic density. It is thought that this elaborate cytoskeletal region is critical for receptor targeting, clustering, and efficient signal input. Gephyrin, a 93 kDa protein, was identified as a result of its ability to bind to polymerized tubulin (microtubules). Although expressed in all tissues, gephyrin mRNA is found most abundantly in brain. Alternative splicing produces at least five different mRNAs. Gephyrin colocalizes and interacts with the glycine receptor at the postsynaptic density. It is possible that Gephyrin acts as an anchor between the glycine receptor and microtubules.
Specifica
| Gephyrin | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Rat Gephyrin aa. 569-726 | |
| 150 μg | |
| Cell Biology | |
| Chicken, Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| 45 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only.
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto