Learn More
Gephyrin Mouse, Unlabeled, Clone: 45, BD
Mouse Monoclonal Antibody
461.17€
Specifica
| Antigene | Gephyrin |
|---|---|
| Clone | 45 |
| Concentrazione | 250μg/mL |
| Applicazioni | Western Blot |
| Classificazione | Monoclonal |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
15865529
|
BD Biosciences
610585 |
150 μg |
462.00€ 461.17€ / 1 set Risparmia 0.83€ 0.18% Sconto |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
The sub-membraneous region of the postsynaptic neuron is an intricate network of cytoskeletal elements generally known as the postsynaptic density. It is thought that this elaborate cytoskeletal region is critical for receptor targeting, clustering, and efficient signal input. Gephyrin, a 93kDa protein, was identified as a result of its ability to bind to polymerized tubulin (microtubules). Although expressed in all tissues, gephyrin mRNA is found most abundantly in brain. Alternative splicing produces at least five different mRNAs. Gephyrin colocalizes and
interacts with the glycine receptor at the postsynaptic density. It is possible that Gephyrin acts as an anchor between the glycine receptor and microtubules.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifica
| Gephyrin | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 45 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Chicken, Human, Mouse, Rat | |
| Rat Gephyrin aa. 569-726 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.