Learn More
Hsp70 Mouse, Unlabeled, Clone: 7, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 610608
Descrizione
Hsp70 is a member of the heat shock or stress-induced genes whose expression is regulated at both transcriptional and translational levels. Elevated temperatures induce a transient resistance to subsequent heat exposure in various types of cells. Although Hsp70 protein is constitutively expressed at low levels, its expression abundantly increases in response to heat stress. During such stress, Hsp70 leaves the cytoplasm and nucleoplasm and rapidly accumulates in the nucleolus. Removal of the stress results in the redistribution of Hsp70 to its original locations.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifica
| Hsp70 | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 150 μg | |
| Cell Biology | |
| Canine, Chicken, Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| 7 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only.