missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFITM3, Human, Matched Antibody Pair (AP62), Abnova™
Marca: Abnova H00010410-AP62.1Set
Dettagli aggiuntivi : Peso : 0.00010kg
Includes: Capture antibody: mouse monoclonal anti-IFITM3 (100μg); Detection antibody: rabbit MaxPab™ affinity purified polyclonal anti-IFITM3 (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Descrizione
Host Species: Mouse, Rabbit
Species Reactivity: Hu
Specifica
IFITM3 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human IFITM3. | |
1-8U/IP15 | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-IFITM3 (100 μg) 2. Detection antibody: rabbit MaxPab™ affinity purified polyclonal anti-IFITM3 (50 μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
IFITM3 | |
IFITM3 | |
Affinity Purified | |
RUO | |
10410 | |
Human |