Learn More
IFN-γ Rat anti-Mouse, APC-Cy7, Clone: XMG1.2, BD
Rat Monoclonal Antibody
Marca: BD Biosciences 561479
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
The XMG1.2 monoclonal antibody specifically binds to mouse interferon- γ(IFN- γ) protein. IFN- γis a pleiotropic cytokine, of approximately 15-17 kDa, involved in the regulation of inflammatory and immune responses. It plays an important role in activation, growth, and differentiation of T and B lymphocytes, macrophages, NK cells and other non-hematopoietic cell types. IFN- γproduction is associated with the Th1 cell differentiation. The purified form of this antibody has been reported to be a neutralizing antibody.
Specifica
IFN-γ | |
Monoclonal | |
0.2mg/mL | |
IFN-γ; IFN-g; IFN-gamma; Interferon gamma; Type II Interferon | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
XMG1.2 | |
APC-Cyanine7 | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Mouse IFN-γ Recombinant Protein | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |