Learn More
NuMA Mouse, Unlabeled, Clone: 22, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 610561
Descrizione
NuMA (Nuclear Mitotic Apparatus protein) is a 2115 amino acid protein with a coiled-coil structure similar to that of myosins and intermediate filaments. Indirect immunofluorescence assays indicate that NuMA's localization is very dynamic. During interphase, NuMA is in the nucleus and during mitosis it moves to the polar regions of the mitotic spindle. NuMA is a very abundant phosphoprotein and antibodies to this protein are often found in patients with autoimmune diseases. Although NuMA is thought to be a structural component of the nucleus, the precise cellular function for this protein is still unknown.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifica
| NuMA | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 50 μg | |
| Cell Biology | |
| Canine, Chicken, Human, Mouse, Rat | |
| IgM |
| Western Blot | |
| 22 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only.