Learn More
NuMA Mouse, Unlabeled, Clone: 22, BD
Mouse Monoclonal Antibody
180.83€ - 401.25€
Specifica
| Antigene | NuMA |
|---|---|
| Clone | 22 |
| Concentrazione | 250μg/mL |
| Applicazioni | Western Blot |
| Classificazione | Monoclonal |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
15865519
|
BD Biosciences
610561 |
50 μg |
181.00€ 180.83€ / 1 set Risparmia 0.17€ 0.09% Sconto |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
15875519
|
BD Biosciences
610562 |
150 μg |
402.00€ 401.25€ / 1 set Risparmia 0.75€ 0.19% Sconto |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
NuMA (Nuclear Mitotic Apparatus protein) is a 2115 amino acid protein with a coiled-coil structure similar to that of myosins and intermediate filaments. Indirect immunofluorescence assays indicate that NuMA's localization is very dynamic. During interphase, NuMA is in the nucleus and during mitosis it moves to the polar regions of the mitotic spindle. NuMA is a very abundant phosphoprotein and antibodies to this protein are often found in patients with autoimmune diseases. Although NuMA is thought to be a structural component of the nucleus, the precise cellular function for this protein is still unknown.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifica
| NuMA | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgM | |
| Affinity Purified |
| 22 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Chicken, Human, Mouse, Rat | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.