missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-PNMA2 Ab-3 Matched Antibody Pair, Abnova™
Marca: Abnova H00010687-AP43.1Set
Dettagli aggiuntivi : Peso : 0.00010kg
Includes: Capture antibody: mouse monoclonal anti-PNMA2 (100μg); Detection antibody: biotinylated mouse monoclonal anti-PNMA2, IgG2b Kappa (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Descrizione
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PNMA2
Host Species: Mouse, Mouse
Isotype: IgG, IgG2b κ
Species Reactivity: Hu
ELISA Pair
Specifica
PNMA2 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PNMA2. | |
KIAA0883/MA2/MM2/RGAG2 | |
Mouse | |
RUO | |
10687 | |
Human | |
Liquid |
ELISA | |
Biotin | |
PNMA2 | |
PNMA2 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-PNMA2 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-PNMA2, IgG2b Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
IgG, IgG2b κ |