missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDE2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA546779
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Peptide sequence: VSAEISENRK RQWPTKSQTD RGASAGKRRC FWLGMEGLET AEGSNSESSD Sequence homology: Human: 100%
SDE2 Polyclonal antibody specifically detects SDE2 in Human samples. It is validated for Western Blot.Specifica
SDE2 | |
Polyclonal | |
Unconjugated | |
SDE2 | |
C1orf55; dJ671D7.1; FLJ35382; protein SDE2 homolog; Replication stress response regulator SDE2; RGD1305572; SDE2; SDE2 telomere maintenance homolog; SDE2 telomere maintenance homolog (S. pombe); UPF0667 protein C1orf55; UPF0667 protein C1orf55 homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
163859 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6IQ49 | |
SDE2 | |
Synthetic peptide directed towards the middle region of human SDE2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |