Learn More
Vimentin Mouse anti-Human, Alexa Fluor 488, Clone: RV202, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 562338
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Intermediate filaments (IF) are a subset of cytoskeletal proteins which function to give overall structural integrity to the plasma membrane as well to organize cells into specific tissues. IF proteins can be divided into six major types based upon the similarity in sequence. Vimentin belongs to the type III category of IF proteins which are expressed in leukocytes, blood vessel endothelial cells, some epithelial cells, and mesenchymal cells. Vimentin is also expressed together with several other IF proteins during the early stages of development. Vimentin is exchanged for the tissue-specific intermediate filament type as differentiation proceeds. Vimentin migrates in SDS/PAGE as a ∽57 kDa protein.
Specifications
Vimentin | |
Monoclonal | |
Alexa Fluor 488 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 |
Flow Cytometry | |
RV202 | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
Purified cow lens vimentin | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |