missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOL4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10564-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene is a member of the apolipoprotein L family and may play a role in lipid exchange and transport throughout the body, as well as in reverse cholesterol transport from peripheral cells to the liver. Two transcript variants encoding two different isoforms have been found for this gene. Only one of the isoforms appears to be a secreted protein.Specifica
APOL4 | |
Polyclonal | |
Unconjugated | |
APOL4 | |
APOL IV, APOL4, Apolipoprotein L IV, apolipoprotein L, 4, Apolipoprotein L4, APOLIV | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80832 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.81 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BPW4 | |
APOL4 | |
APOL4 Fusion Protein Ag0849 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |