Learn More
SI Analytics™ AquaLine Electrode
SI Analytics™ AquaLine Electrode is a low maintenance electrode for drinking water range.
Marca: SI Analytics™ 285113049
Dettagli aggiuntivi : Peso : 0.25000kg
Descrizione
- AquaLine electrodes for drinking water range
- Low maintenance
- Included a screw plug head
- pH combination electrode
- Glass shaft
- Ceramic junction
- Gel electrolyte
- Conical membrane
Specifica
Plug Head | |
2 to 13 | |
Silamid™ | |
Aqualine 70 PH | |
12 mm |
Glass | |
pH Combination Electrode | |
Cone | |
120 mm |