missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin 8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA561197
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: EIAMCEPEFG NDKAREPSVG GRWRVSWYER F Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 35%.
Aquaporin-8 is a protein that in humans is encoded by the AQP8 gene. Aquaporin 8 (AQP8) is a water channel protein. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). Aquaporin 8 mRNA is found in pancreas and colon but not other tissues. Forms a water-specific channel; mercury-sensitive. Not permeable to glycerol or urea. Expressed only in pancreas and colon.Specifica
Aquaporin 8 | |
Polyclonal | |
Unconjugated | |
Aqp8 | |
AI255744; AQP8; AQP-8; aquaporin 8; Aquaporin8; aquaporin-8; membrane water channel; MGC93492; water channel | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
343 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94778 | |
Aqp8 | |
Recombinant protein corresponding to Human Aquaporin 8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only