missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ AR (Human) Recombinant Protein
Human AR partial ORF ( NP_000035, 221 a.a. - 320 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00000367-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
- Sequence: SKDNYLGGTSTISDNAKELCKAVSVSMGLGVEALEHLSPGEQLRGDCMYAPLLGVPPAVRPTPCAPLAECKGSLLDDSAGKSTEDTAEYSPFKGGYTKGL
Specifica
NP_000035 | |
androgen receptor | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AR | |
GST |
367 | |
Wheat germ expression system | |
25 μg | |
AIS, DHTR, HUMARA, KD, NR3C4, SBMA, SMAX1, TFM | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |