missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARID1B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA550918
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
ARID1B / BAF250B Polyclonal Antibody detects endogenous levels of ARID1B / BAF250B protein.
ARID1B Polyclonal antibody specifically detects ARID1B in Human, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot.Specifica
ARID1B | |
Polyclonal | |
Unconjugated | |
ARID1B | |
6A3-5; 8030481M12; 9330189K18Rik; AI836955; Ardi1b; ARID domain-containing protein 1B; ARID1B; AT rich interactive domain 1B (Swi1 like); AT rich interactive domain 1B (SWI1-like); AT rich interactive domain 1B (SWI-like); AT-rich interaction domain 1B; AT-rich interactive domain-containing protein 1B; B230217J03Rik; BAF250B; BRG1-associated factor 250b; BRG1-binding protein ELD/OSA1; BRG1-binding protein hELD/OSA1; BRIGHT; CSS1; DAN15; ELD (eyelid)/OSA protein; ELD/OSA1; hOsa2; KIAA1235; mKIAA1235; MRD12; Osa homolog 2; OSA2; P250R; Smcf1; smcf-1; transcription factor 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
282546, 57492 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8NFD5 | |
ARID1B | |
Synthesized peptide derived from the Internal region of human BAF250b. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |