Learn More
ARMCX5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA551496
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: QAKAGDGAMT RTHTVTYREA MAVTREVIKV EDTTKTRVMV ETKTKPLAER SIVPQTKSKA MPMSRVSTVT KSEVKVVAVI EANIRSYAKS HDKANTGSRP DRREETSIGM KSSDEDEE Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 41%.
ARMCX5 is a protein coding gene.Specifica
ARMCX5 | |
Polyclonal | |
Unconjugated | |
ARMCX5 | |
armadillo repeat containing, X-linked 5; armadillo repeat-containing X-linked protein 5; ARMCX5; GASP5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64860 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6P1M9 | |
ARMCX5 | |
Recombinant protein corresponding to Human ARMCX5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only