Learn More
ARPIN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA562582
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: EVMELELGAG VRLKTRGDGP FLDSLAKLEA GTVTKCNFTG DGKTGASWTD NIMAQKCSKG AAAEIREQGD GAEDEEWDD Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
ARPIN gene ontology annotations related to this gene include negative regulation of actin nucleation.Specifica
ARPIN | |
Polyclonal | |
Unconjugated | |
ARPIN | |
2610034B18Rik; actin related protein 2/3 complex inhibitor; actin-related protein 2/3 complex inhibitor; arp2/3 inhibition protein; Arpin; C15orf38; C21H15orf38; C3H15orf38; C7H15orf38; hypothetical protein LOC550477; RGD1311021; RIKEN cDNA 2610034B18 gene; si:ch211-200o3.3; UPF0552 protein C15orf38; UPF0552 protein C15orf38 homolog; zgc:112432 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
348110 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z6K5 | |
ARPIN | |
Recombinant protein corresponding to Human ARPIN | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only