missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASCC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12301-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a subunit of the activating signal cointegrator 1 (ASC-1) complex. The ASC-1 complex is a transcriptional coactivator that plays an important role in gene transactivation by multiple transcription factors including activating protein 1 (AP-1), nuclear factor kappa-B (NF-kB) and serum response factor (SRF). The encoded protein contains an N-terminal KH-type RNA-binding motif which is required for AP-1 transactivation by the ASC-1 complex. Mutations in this gene are associated with Barrett esophagus and esophageal adenocarcinoma. Alternatively spliced transcripts encoding multiple isoforms have been observed for this gene.Specifica
ASCC1 | |
Polyclonal | |
Unconjugated | |
ASCC1 | |
ASC 1 complex subunit p50, ASC1p50, ASCC1, CGI 18, RP11 150D20.4, Trip4 complex subunit p50 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
294512, 51008, 69090 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8N9N2, Q9D8Z1 | |
ASCC1 | |
ASCC1 Fusion Protein Ag2946 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |