Learn More
ATAGO™ RX-9000α Automatic Digital Refractometer
Designed with a wide measurement range, measurement temperature range, and high measurement accuracy, the ATAGO™ RX-9000α Automatic Digital Refractometer is suitable for multiple sample types such as measuring oils and fats with high melting points, aroma oils of high refractive index and organic solvents, meeting demands for high accuracy measurements.
Marca: ATAGO™ 3263
Descrizione
- Fully automatic digital refractometer with high level of accuracy
- Built-in Peltier thermo module for maintaining a constant sample temperature while taking measurements
- Manual calibration is also possible to adjust to standard liquid values and to correct differences in measurement values between instruments
Specifica
Refractometer | |
1.32500 to 1.70000 | |
Refrac index (nD): ±0.00004 (nD 1.33299 to1.42009 at 10 to 30°C ); Refrac index (nD): ±0.00010 (For ranges other than above); Brix ±0.03% (Brix 0.00 to 50.00% at 10 to 30°C ); Brix ±0.05% (Brix 50.01 to 95.00% at 10 to 30°C ); Brix ±0.10% (For ranges other than above, with Sucrose solution by MODE-1) | |
Min. Indication: Refractive index (nD): 0.00001; Brix 0.01% | |
5°C to 70°C | |
0.01°C |
0.00 to 100.00 Brix | |
RX-9000α | |
Refrac index (nD) ±0.00002 (nD 1.33299 to 1.42009 at 10 to 30°C ); Refrac index (nD): ±0.00005 (For ranges other than above); Brix ±0.01% (Brix 0.00 to 50.00% at 10 to 30°C ); Brix ±0.02% (For ranges other than above, with Sucrose solution by MODE-1) | |
Brix, Refractive Index | |
5°C to 70°C |