missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aurora A Antibody (1A11), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-50040SS
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Aurora A Monoclonal antibody specifically detects Aurora A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifica
| Aurora A | |
| Monoclonal | |
| Unconjugated | |
| Supplied as concentrated hybridoma cell culture media. | |
| AIKAurA, ARK-1, ARK1EC 2.7.11.1, AURA, aurora kinase AhARK1, aurora/IPL1-like kinase, Aurora/IPL1-related kinase 1, Aurora-related kinase 1, Breast tumor-amplified kinase, breast-tumor-amplified kinase, BTAKAURORA2, EC 2.7.11, IPL1-related kinase, serine/threonine kinase 6, serine/threonine protein kinase 15, Serine/threonine-protein kinase 15, serine/threonine-protein kinase 6, Serine/threonine-protein kinase aurora-A, STK15serine/threonine kinase 15, STK6MGC34538, STK7 | |
| Full length recombinant human protein aurora A expressed in and purified from E. coli | |
| 0.125 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, MAP Kinase Signaling, Mitotic Regulators, Protein Kinase, Signal Transduction, Stem Cell Markers, Tumor Suppressors | |
| 6790 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 1A11 | |
| Western Blot 1:100 - 1:500, Immunohistochemistry 1:100-1:500, Immunocytochemistry/ Immunofluorescence 1:100 - 1:500 | |
| O14965, O14965 | |
| Mouse | |
| Tissue culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Supernatant |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto