Learn More
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm
Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency.
Marca: Thermo Scientific™ 908-262
315.60 EUR valido fino al 2024-12-31
Usa il codice promo "21898" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 9.44000kg
Descrizione
Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Tecan Te-MO, Aquarius and Freedom EVO with Te-MO 96 head.
- Available nonsterile with-or-without Thermo Scientific™ ART™ filter technology
- Available in filtered, low retention or extended length for increased volume to reach into deep well plates
- Lot-tested during production on actual workstation for which they are created
- Fully conductive providing increased pipetting performance
- Low percentage coefficient of variation (%CV)
Specifica
200 μL | |
Non-sterile | |
Rack | |
Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius | |
Certified Quality |
Non-Filtered | |
Clear | |
Low Retention, Extended Length | |
Tecan |