Learn More
Novacyt - Microgen Bioproducts Avian Allergens Serological Double Gel Diffusion Assay
Microgen Bioproducts Avian Allergens - A serological double gel diffusion assay intended for the detection of precipitating antibodies to avian allergens
Marca: Novacyt - Microgen Bioproducts FSK4
Dettagli aggiuntivi : Peso : 0.50000kg
Descrizione
- Double diffusion assay for the detection of precipitating antibodies elicited to the inhalation of avian allergens.
Specifica
6.5 mL | |
Double gel diffusion (Ouchterlony) | |
Test Kit |
Liquid | |
Double Gel Diffusion Assay | |
5 to 10 mL of venous blood |