Learn More
Axygen™ Axypet™ Single Channel Pipettor, Calibration 3x10 STD
High levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods of time.
Marca: Axygen™ AP-20-STD
267.66 EUR valido fino al 2024-12-31
Usa il codice promo "23272" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.23080kg
Descrizione
- ISO 17025 Pre-calibrated
- Thumb-friendly soft spring pipetting system reduces fatigue
- Unique side EZIject button option reduces thumb movement (Left handed EZIject button available by special order).
- Finger-hook provides for a more relaxed grip
- Large ergonomic push button is easy on the thumb during contact
- Locking feature prevents accidental volume changes during pipetting procedures
Specifica
ISO17025 | |
Axypet™ | |
2 to 20 μL |
1 | |
Single Channel Pipettor | |
1/Pk., 1/Cs |