missing translation for 'onlineSavingsMsg'
Learn More

BCAS2 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10414-1-AP-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16896783

  • 325.38€ / 150µL

Spedizione stimata: 06-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

cDNA encoding BCAS2 has been isolated. The deduced 225-amino acid protein contains a central potential N-glycosylation site and multiple N-terminal potential phosphorylation sites. Northern and Southern blot analyses demonstrates expression of a 1. 5-kb transcript at various levels in multiple breast cancer cell lines, with greatest amplification in BT-20 and MCF-7 cells. BCAS2 is ubiquitously expressed in normal tissues.
Specifica

Specifica

BCAS2
Polyclonal
Unconjugated
BCAS2
BCAS2, DAM1, Pre mRNA splicing factor SPF27
Rabbit
Antigen Affinity Chromatography
RUO
10286, 295334, 68183
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.34 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O75934, Q9D287
BCAS2
BCAS2 Fusion Protein Ag0669
150 μL
Primary
Rat, Non-human Primate, Mouse, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali