Learn More
BCAS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10402-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
BCAS3 is physically associated with histone H3 and histone acetyltransferase complex protein P/CAF (p300/CBP-associated factor) and possesses histone acetyltransferase activity. In breast cancer, several chromosomal sites frequently undergo amplification, implicating the location of genes important for tumor development and progression.Specifica
| BCAS3 | |
| Polyclonal | |
| Unconjugated | |
| BCAS3 | |
| BCAS3, GAOB1, MAAB, Protein Maab1, Rudhira | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 192197, 363662, 54828 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8CCN5, Q9H6U6 | |
| Bcas3 | |
| BCAS3 Fusion Protein Ag0646 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |