Learn More
Abnova™ BCL2 Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Marca: Abnova™ FS0052.100uL
Dettagli aggiuntivi : Peso : 0.02000kg
Specifica
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
Bcl-2 | |
596 | |
100 μL |
Tube | |
B-cell CLL/lymphoma 2 | |
BCL2 | |
DNA/RNA | |
Liquid |