missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL2L10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18114-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. The protein encoded by this gene contains conserved BH4, BH1 and BH2 domains. This protein can interact with other members of BCL-2 protein family including BCL2, BCL2L1/BCL-X(L), and BAX. Overexpression of this gene has been shown to suppress cell apoptosis possibly through the prevention of cytochrome C release from the mitochondria, and thus activating caspase-3 activation. The mouse counterpart of this protein is found to interact with Apaf1 and forms a protein complex with Caspase 9, which suggests the involvement of this protein in APAF1 and CASPASE 9 related apoptotic pathway.Specifica
BCL2L10 | |
Polyclonal | |
Unconjugated | |
BCL2L10 | |
Anti apoptotic protein NrH, Apoptosis regulator Bcl B, BCL2L10, Boo, Diva | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10017 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9HD36 | |
BCL2L10 | |
BCL2L10 Fusion Protein Ag12596 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |