missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Soybean Casein Digest Agar
Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms
Marca: BD 211046
Dettagli aggiuntivi : Peso : 2.48000kg
Descrizione
Applications:
Isolation of non-fastidious and fastidious
Specifica
ISO/DIS, 11014, AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP | |
Dehydrated | |
Soybean Casein Digest Agar | |
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean 5g/L, Sodium Chloride 5g/L, Agar 15g/L | |
Pail |
25°C | |
Soy Agar | |
Soybean Casein digest Agar | |
Isolation of non-fastidious and fastidious | |
2.3 kg |