Learn More
BD BV480 Rat Anti-Mouse CD45
BD Horizon - BV480 Rat Anti-Mouse CD45 - CD45 is a transmembrane glycoprotein which is expressed at high levels on cell-surface, and its presence distinguishes leukocytes from non-hematopoietic cells. CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction and the CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific.
Marca: BD 566095
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon BV480 can be excited by the violet laser and detected in the BD Horizon BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifica
BV480 Rat Anti-Mouse CD45 | |
Flow Cytometry | |
QC Testing: Mouse | |
Store undiluted at 4°C |
50 μg | |
0.2 mg/mL | |
BV480 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |