missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD OptEIA™ Mouse IL-3 ELISA Set
Develop ELISAs for natural or recombinant mouse IL-3 in serum, plasma and cell culture supernatants.
Marca: BD 555228
Dettagli aggiuntivi : Peso : 0.16000kg
Includes:
- Capture Antibody: Anti-mouse IL-3
- Detection Antibody: Biotinylated anti-mouse IL-3
- Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP)
- Standard: Recombinant mouse IL-3 (lyophilized)
Descrizione
- Sufficient materials are provided to yield approximately 20 plates of 96-wells
Enzyme-linked immunosorbent assays (ELISA)
Specifica
ELISA Kit | |
Mouse | |
Liquid | |
Capture Antibody, detection Antibody, Streptavidin-HRP, standards |
20 Plates | |
Mouse | |
ELISA | |
4°C |