missing translation for 'onlineSavingsMsg'
Learn More
Learn More
behr Labor-Technik™ behrotest™ Process Extraction System,behrosog neutralizer
Obtain precise extraction with this process extraction system. behr Labor-Technik™ behrotest™ Process Extraction System is perfect for vacuuming and washing aggressive acid vapors with a vacuum pump.
Marca: behr Labor-Technik™ B00217925
Descrizione
- The two-level behrosog process extraction system (neutralizer) – pre-separator plus safety level – keeps all acid vapors entirely away from the environment
- Compact process extraction system for extracting and neutralizing aggressive acid vapors, especially from the Kjeldahl digestion for nitrogen determination.
- An upstream two-level pre-separator washes the toxic substance out. The process extraction system is equipped with a vacuum pump 40 l/min.
- There is entirely no need for a connection to the water supply.
- Optionally, the additional cooling system ACS for behrosog 3, consisting of stand, bottle and cooler, can be connected to the system.
- Efficient operating costs
- Infinitely regulated extraction through-flow
- Both cleaning levels (condensation and neutralization level) prevent the emission of toxic substances into the environment
- extremely environmental friendly
- No connection to water supply needed
- infinite regulation of air flow quantity 16 ... 40 l/min
- easy exchange of liquids through simply emptying of glass vessels
- PTFE-coated double-head membrane pump
- Integrated droplet separator
- Gas wash bottle 2000 ml for cooling hot gases
- Gas wash bottle 2000 ml for neutralizing
- On/off switch outside hazard zone
- Transparent safety door
- Removable safety drip tray
- Removable safety drip tray
- Transparent safety door
- On/off switch outside hazard zone
- Gas wash bottle 2000 ml for neutralizing
- Gas wash bottle 2000 ml for cooling hot gases
- Integrated droplet separator
- PTFE-coated double-head membrane pump
- easy exchange of liquids through simply emptying of glass vessels
- infinite regulation of air flow quantity 16 ... 40 l/min
- No connection to water supply needed
- extremely environmental friendly
- Both cleaning levels (condensation and neutralization level) prevent the emission of toxic substances into the environment
- Infinitely regulated extraction through-flow
- Efficient operating costs
- Optionally, the additional cooling system ACS for behrosog 3, consisting of stand, bottle and cooler, can be connected to the system.
- There is entirely no need for a connection to the water supply.
- An upstream two-level pre-separator washes the toxic substance out. The process extraction system is equipped with a vacuum pump 40 l/min.
- Compact process extraction system for extracting and neutralizing aggressive acid vapors, especially from the Kjeldahl digestion for nitrogen determination.
Specifica
40 L/hr | |
0.41 m | |
0.41 m |
Scrubber with suction pump (40 L/min), cooling level, neutralization level and drip catcher | |
0.53 m |