Learn More
Köttermann Bench-type Hood
Designed for universal use, this Bench-type Hood is ideal for dealing with organic solvents and is also suitable for working with cold or diluted acids.
Marca: Köttermann 451-00032
Dettagli aggiuntivi : Peso : 73.35900kg
Descrizione
- All-purpose hood for general use
- Optimized protection from volatile hazardous materials
- Long-lasting steel construction
- Ergonomic features to improve workflow
Specifica
No Ventilation Fan | |
GB Plug | |
85 kg |
800 x 1350 x 640 mm | |
230 V |