Learn More
Emma Safety Footwear Bergamo Safety Shoes
Comfortable foot-hugging fit provides a naturally light and airy foot feel. Virtually penetration proof, water resistant uppers. The 100% nitrile rubber outsole repels liquids, harmful or otherwise and the ingeniously designed sole contours provide extra grip on almost all surfaces.
Marca: Emma Safety Footwear 107092-41
Dettagli aggiuntivi : Peso : 1.43900kg
Descrizione
- O3-Work shoe without steel toecap with sole plate
- Full grain leather construction
- Nitrile Rubber Heat Resistant Outsole
- Metal-free
- Chrome-free lining
- ESD certified
- Sanitized
Specifica
EN ISO 20347:2012 | |
Black | |
Hydro-Tec™ Sanitized Silver & Wellness leather | |
41 |
Laces | |
Bergamo | |
Leather |