Learn More
beta-2 Adrenergic R/ADRB2 Antibody (4C11), Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP3-26521-50ul
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
beta-2 Adrenergic R/ADRB2 Monoclonal antibody specifically detects beta-2 Adrenergic R/ADRB2 in Human samples. It is validated for ELISA, Immunohistochemistry
Specifica
| beta-2 Adrenergic R/ADRB2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| 4C11 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| ADRBR, adrenergic, beta-2-, receptor, surface, B2ARADRB2R, BAR, beta-2 adrenergic receptor, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor | |
| A synthesized peptide derived from Human beta-2 Adrenergic R/ADRB2 [UniProt P07550] | |
| 50 μL | |
| Asthma, GPCR, Immunology, Neuroscience, Phospho Specific, Signal Transduction | |
| 154 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |