missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-2-microglobulin Mouse anti-Human, Clone: 1C3B7, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66207-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Beta-2-microglobulin (B2M) is a 12-14 kDa protein that is a subunit of the major histocompatibility complex (MHC) class I. Human beta 2 Microglobulin associated with HLA Class I antigens are expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells, but is absent on erythrocytes. Beta-2 microglobulin is also found in free form in the plasma. The amino acid sequence of Beta-2-microglobulin is homologous to the constant regions of immunoglobulin heavy and light chains, with a predomitly beta pleated sheet structure. The protein is ubiquitously expressed in the cytoplasm, and is involved in antigen processing and presentation. Beta-2-microglobulin also plays a role in TCR signaling, and downstream signaling in naive CD8+ cells. Beta 2-microglobulin is encoded by the B2M gene located on chromosome 15. A mutation in the Beta-2-microglobulin gene has been shown to result in hypercatabolic hypoproteinemia. Levels of beta 2 microglobulin can be used to assess renal function in kidney transplant recipients. Beta-2-microglobulin that protein can be used as a prognostic marker for HIV infection.Specifica
Beta-2-microglobulin | |
Monoclonal | |
1.46 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P61769 | |
B2M | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
1C3B7 | |
Unconjugated | |
B2M | |
B2M, Beta-2-microglobulin, HDCMA22P | |
Mouse | |
Protein A | |
RUO | |
567 | |
-20°C | |
Liquid |