Learn More
beta Galactosidase Antibody (BG-02), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB500-334MFV500
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
beta Galactosidase Monoclonal antibody specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifica
| beta Galactosidase | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| EC 3.2.1.23; b-gal | |
| beta-Galactosidase purified from E. coli. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| BG-02 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| E. coli | |
| Purified |