Learn More
Fisherbrand™ Beta Shields
Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Include:
15° Angle
45° Angle
Dimensione della confezione:
Pezzo
Descrizione
- Beta Shield - Small Fixed 15° Angle - Curved Base 30 x 45cm
- Supplied in small, medium and large sizes and with a 15cm-deep curved base for use with safety trays, or a 30x30cm flat base for under-the-bench protection
- A range of angles offers increased manoeuvrability, while clear optical acrylic aids visualisation
- Radiation shielding Cleaver Scientific for protection from Beta radiation, shield, fixed 15 degree angle, curved base, dimensions 450h x 300w x 150d mm
Specifica
Specifica
| Include | 15° Angle |
| Product Type | Beta Shield |