Learn More
Novacyt - Microgen Bioproducts Biochemical Identification System: Listeria spp.
Biochemical identification system for Listeria spp. from foods and other environmental samples.
Marca: Novacyt - Microgen Bioproducts MID-67
Dettagli aggiuntivi : Peso : 1.90000kg
Descrizione
- Twelve-substrate biochemical identification system.
- From a range of different technologies for the specific detection of Listeria spp. from foods and other environmental samples.
- From a range of simple, reliable, bacterial identification systems for food, environmental, and industrial laboratories.
- These tests will arm professionals with the necessary information to maximize the effectiveness of appropriate patient management.
- The range is expanding quickly and will encompass the widest range of microbiological testing solutions.
Specifica
Identification of Listeria Spp. | |
20 Tests |