Learn More
Biochrom Print Via Computer (PVC) Software with USB Cable
Print Via Computer (PVC) Software with USB Cable enables Biochrom spectrophotometer users to print through the PC directly to the printer connected to it. Data can also be stored in Excel, EMF, comma delimited (csv), tab delimited (txt) data files or native PVC format.
Marca: Biochrom 80300473
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Supplied with USB cable
- Compatible with Windows 2000 - Windows XP
- Prints to ANY printer connected to PC (or networked )
- Up to 8 instruments may be configured to 1 PC (ports allowing)
- Displays results on PC
- Multiple File formats supported including PVC format, Excel Format, Windows Enhanced Metafile (graphic file) and .TXT, .CSV ASCII formats
Specifica
Windows 2000 - Windows XP | |
Print Via Computer Software | |
USB cable |