missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Falcon™ Biocoat™ 384-Well, Collagen Type I-Treated, Flat-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:With Lid
Marca: Falcon™ 356667
Dettagli aggiuntivi : Peso : 3.60000kg
Descrizione
- Source: Rat tail
- TC-treated
- Clear, round well
- Flat bottom
- Non-sterile
- Promotes cell growth, differentiation
- Cell adhesion assays, tissue morphogenesis, cell-matrix interaction, receptor-ligand binding
Specifica
Standard | |
4°C to 30°C (Stable for 12 months) | |
Black, Clear Bottom | |
Non-sterile | |
Flat Bottom (F-well) | |
50/Cs. |
With Lid | |
16 x 24 | |
384 | |
Collagen Type I | |
Attachment and spreading of HT-1080 human fibrosarcoma cells | |
Rat tail collagen |