Learn More
ACROBiosystems Biotinylated Cynomolgus / Rhesus macaque BCMA / TNFRSF17 Protein, His, Avitag™
Biotinylated Protein
Marca:  ACROBiosystems BCA-C82E5-200ug
Dettagli aggiuntivi : Peso : 0.00010kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17106091
  • dietro preventivo / 200µg
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Descrizione
Descrizione
Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 9.6 kDa. The protein migrates as 15-18 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifica
Specifica
XP_005591343.1
9.6 kDa
200 µg
Met 1 - Ala 53
TNFRSF17,CD269,BCM,BCMA
Precision Avi
PBS, pH7.4
HEK293
Cynomolgus / Rhesus macaque
Powder
>90% as determined by SDS-PAGE
Biotinylated Cynomolgus / Rhesus macaque BCMA, His,Avitag (BCA-C82E5) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession # G7Q0I4-1).
-20°C
BCMA
His Tag and Avi Tag
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins