Learn More
ACROBiosystems Biotinylated Human FGL1 (64-312) Protein, Avitag™,Fc Tag (high sensitivity)
Biotinylated Protein
Marca:  ACROBiosystems FG1-H82F6-25ug
Dettagli aggiuntivi : Peso : 0.00010kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17116321
  • dietro preventivo / 25µg
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Descrizione
Descrizione
Fibrinogen-like protein 1(FGL1) is also known as HP-041, Hepassocin, HFREP-1, LFIRE-1. The protective effect of fibrinogen-like protein 1 (FGL1) in liver injury has previously been reported. However, studies have shown that FGL1 may be a predictor of GC patients and a target for GC therapy. Immunocytochemical studies revealed that fgl1 selectively binds to defective spermatozoa in the cauda epididymidis. Northern blot analysis and in situ hybridization demonstrated the high expression of fgl1 in the principal cells of the proximal cauda epididymidis. Immunofluorescence analysis using mouse fibrotic lung tissues suggested that fibrotic regions showed increased expressions of Gtse1 and Fgl1, Gtse1 and Fgl1 are suggested to be novel targets for radiation-induced lung fibrosis.
This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 116.3 kDa. The protein migrates as 110-120 kDa under reducing (R) condition (SDS-PAGE).
Specifica
Specifica
NP_004458.3
116.3 kDa
25 µg
Asp 64 - Ile 312
FGL1,Hepassocin,HP-041,HFREP-1,LFIRE-1,HFREP1
Precision Avi
50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5
HEK293
Human
Powder
>90% as determined by SDS-PAGE
Biotinylated Human FGL1 (64-312), Avitag,Fc Tag (FG1-H82F6) is expressed from human 293 cells (HEK293). It contains AA Asp 64 - Ile 312 (Accession # Q08830-1).
-20°C
FGL1
Avi Tag and Fc Tag
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins