Learn More
ACROBiosystems Biotinylated Human Latent TGF-beta 3 / Latent TGFB3 Protein, Avitag™
Biotinylated Protein
Marca:  ACROBiosystems TG3-H8218-200ug
Dettagli aggiuntivi : Peso : 0.00010kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17106831
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Descrizione
Descrizione
Transforming growth factor beta 3 ( TGFB3) is also known as TGF-β3, is a polypeptide member of the transforming growth factor beta superfamily of cytokines. It is a secreted protein that performs many cellular functions, including regulates embryogenesis and cell differentiation and is required in various processes such as secondary palate development. TGF-beta 3 is similar with TGF-beta 1 and ­-beta 2, act as cellular switches to regulate immune function, cell proliferation, and epithelial­-mesenchymal transition. TGF-beta-3 is released from LAP by integrins: integrin-binding results in distortion of the LAP chain and subsequent release of the active TGF-beta-3. Once activated following release of LAP, TGF-beta-3 acts by binding to TGF-beta receptors (TGFBR1 and TGFBR2), which transduce signal.
This protein carries an Avi tag at the C-terminus. The protein has a calculated MW of 14.4 kDa.
Specifica
Specifica
Powder
200 µg
Ala 301 - Ser 412
TGFB3,ARVD,TGF-beta3
Unconjugated
HEK293
Human
14.4 kDa
Biotinylated Human Latent TGFB3, Avitag (TG3-H8218) is expressed from human 293 cells (HEK293). It contains AA Ala 301 - Ser 412 (Accession # P10600-1).
-20°C
TGF-beta 3
Avi Tag
MABSol™ Biotin Labeled Proteins