Learn More
ACROBiosystems Biotinylated Human TNFSF11 / RANKL / CD254 Protein, His,Avitag™, active trimer
MALS verified
Marca:  ACROBiosystems RAL-H82Q5-200ug
Dettagli aggiuntivi : Peso : 0.00010kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17106651
  • 2103.34€ / 200µg
Spedizione stimata: 15-05-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Receptor activator of nuclear factor kappa-B ligand (RANKL), also known as tumor necrosis factor ligand superfamily member 11 (TNFSF11), TNF-related activation-induced cytokine (TRANCE), osteoprotegerin ligand (OPGL), and osteoclast differentiation factor (ODF), is known as a type II membrane protein and is a member of the tumor necrosis factor (TNF) superfamily. RANKL, through its ability to stimulate osteoclast formation and activity, is a critical mediator of bone resorption and overall bone density. Some findings also suggestion some cancer cells, particularly prostate cancer cells, can activate an increase in bone remodeling and ultimately increase overall bone production.[17] This increase in bone remodeling and bone production increases the overall growth of bone metastasizes. The overall control of bone remodeling is regulated by the binding of RANKL with its receptor or its decoy receptor, respectively, RANK and OPG.
This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 24.1 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifica
Specifica
AAC51762.1
200 μg
Gly 64 - Asp 245
RANKL,CD254,TRANCE,OPGL,ODF
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
24.1 kDa
Biotinylated Human TNFSF11, His,Avitag, active trimer (RAL-H82Q5) is expressed from human 293 cells (HEK293). It contains AA Gly 64 - Asp 245 (Accession # AAC51762.1) trimer Design.
-20°C
TNFSF11
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins