Learn More
ACROBiosystems Biotinylated Mouse IL-6 R alpha / CD126 Protein, His, Avitag
Marca:  ACROBiosystems ILR-M82E9-200ug
Dettagli aggiuntivi : Peso : 0.00010kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17236280
  • dietro preventivo / 200µg
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Descrizione
Descrizione
Interleukin 6 receptor (IL6R) is also known as CD126 (Cluster of Differentiation 126) , is a potent pleiotropic cytokine that regulates cell growth and differentiation of various tissues, and is known particularly for its role in the immune response and acute phase reactions. IL6R is a protein complex consisting of a IL-6 receptor subunit (IL6R) and interleukin 6 signal transducer Glycoprotein 130. IL6R also denotes the human gene encoding this subunit. Alternatively spliced transcript variants encoding distinct isoforms have been reported. IL6R subunit also shared by many other cytokines. The soluble form of IL6R arises from proteolytic cleavage of membrane-bound IL6Rα, and acts agonistically by making the IL6 ligand accessible to the signal transducer gp130. Dysregulated production of IL6 and IL6R are implicated in the pathogenesis of several inflammatory diseases and malignancies such as multiple myeloma, rheumatoid arthritis, or osteoporosis, and it has been reported that a humanized anti-IL6R monoclonal antibody is a promising agent applicable to the therapeutic approach for IL6 driven diseases. Interleukin-6 receptor has been shown to interact with Interleukin 6 and Ciliary neurotrophic factor.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 41.0 kDa. The protein migrates as 45-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifica
Specifica
NP_034689.2
>95% as determined by SDS-PAGE.
Biotinylated Mouse IL-6 R alpha, His,Avitag (ILR-M82E9) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Glu 357 (Accession # P22272-1).
-20°C
IL-6 R alpha
His Tag & Avi Tag
Less than 1.0 EU / µg by the LAL method.
Powder
Mouse
41.0 kDa
200 µg
Leu 20 - Glu 357
IL6R,CD126,IL-6 R-1,IL-6 RA,IL6RA,IL-6 R-alpha,IL6RQ,gp80
Precision Avi
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins